Methyl-5-[(4-chlorophenyl)sulfanyl]-4-formyl-1-phenyl-1H-pyrazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

043246-500MG 043246-1G 043246-5G
101859-868EA 747.69 USD
101859-868 101859-870 101859-872
Methyl-5-[(4-chlorophenyl)sulfanyl]-4-formyl-1-phenyl-1H-pyrazole-3-carboxylate ≥95%
Methyl-5-[(4-chlorophenyl)sulfanyl]-4-formyl-1-phenyl-1H-pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₁₈H₁₃ClN₂O₃S
Melting Pt: 109…112 °C
MDL Number: MFCD05021848
CAS Number: 449778-53-4

Order Now

Specification Test Results

Melting point 109-112°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR