Ethyl-4,6-dioxo-5-phenyl-1,3a,4,5,6,6a-hexahydropyrrolo[3,4-c]pyrazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

043242-500MG 043242-1G 043242-5G
101859-852EA 1109.23 USD
101859-852 101859-854 101859-856
Ethyl-4,6-dioxo-5-phenyl-1,3a,4,5,6,6a-hexahydropyrrolo[3,4-c]pyrazole-3-carboxylate ≥95%
Ethyl-4,6-dioxo-5-phenyl-1,3a,4,5,6,6a-hexahydropyrrolo[3,4-c]pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₃N₃O₄
Melting Pt: 205…207 °C
MDL Number: MFCD00687177
CAS Number: 2997-63-9

Order Now

Specification Test Results

Melting point 205-207°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR