2-(4-Nonylphenoxy)acetic acid ≥95%

Supplier: Matrix Scientific

043241-500MG 043241-1G
101859-848EA 921.54 USD
101859-848 101859-850
2-(4-Nonylphenoxy)acetic acid ≥95%
2-(4-Nonylphenoxy)acetic acid

Hazard: Irritant

Formula: C₁₇H₂₆O₃
Melting Pt: 95…97 °C
MDL Number: MFCD00016818
CAS Number: 3115-49-9

Order Now

Specification Test Results

Melting point 95-97°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR