4-(1-Methyl-2-oxopropoxy)benzenecarbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
4-[(3-Oxobutan-2-yl)oxy]benzonitrile
043228-500MG
043228-1G
043228-5G
101859-788EA
63.08
USD
101859-788
101859-790
101859-792
4-(1-Methyl-2-oxopropoxy)benzenecarbonitrile ≥95%
4-(1-Methyl-2-oxopropoxy)benzenecarbonitrile
Hazard: Irritant
Formula:
C₁₁H₁₁NO₂ MW: 189.22 g/mol Melting Pt: 74…76 °C |
MDL Number:
MFCD00243580 CAS Number: 306979-60-2 |
Specification Test Results
Melting point | 74-76°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...