2-(Ammoniooxy)propane chloride ≥95%

Supplier: Matrix Scientific

043224-1G 043224-5G
101859-768EA 444.62 USD
101859-768 101859-770
2-(Ammoniooxy)propane chloride ≥95%
2-(Ammoniooxy)propane chloride

Hazard: Irritant

Formula: C₃H₁₀ClNO
Melting Pt: 94-96°C
MDL Number: MFCD00231457
CAS Number: 4490-81-7

Order Now

Specification Test Results

Melting point 94-96°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR