1-(Ammoniooxy)propane chloride ≥95%

Supplier: Matrix Scientific

043223-500MG 043223-1G 043223-5G
101859-760EA 269.23 USD
101859-760 101859-762 101859-764
1-(Ammoniooxy)propane chloride ≥95%
1-(Ammoniooxy)propane chloride

Hazard: Irritant/Refrigerate

Formula: C₃H₁₀ClNO
Melting Pt: 148-152°C
MDL Number: MFCD00231456
CAS Number: 6084-54-4

Order Now

Specification Test Results

Melting point 148-152°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR