4-(4-Methoxyphenyl)-3-buten-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(P-Methoxyphenyl)-1-buten-3-one

043222-500MG 043222-1G 043222-5G
101859-754EA 146.15 USD
101859-754 101859-756 101859-758
4-(4-Methoxyphenyl)-3-buten-2-one ≥95%
4-(4-Methoxyphenyl)-3-buten-2-one

Hazard: Irritant

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Melting Pt: 75…77 °C
MDL Number: MFCD00017251
CAS Number: 943-88-4

Order Now

Specification Test Results

Melting point 75-77°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR