2-(4-Bromophenylsulfonamido)acetic acid ≥95%

Supplier: Matrix Scientific

043220-500MG 043220-1G 043220-5G
101859-744EA 181.54 USD
101859-744 101859-746 101859-748
2-(4-Bromophenylsulfonamido)acetic acid ≥95%
2-(4-Bromophenylsulphonamido)acetic acid

Hazard: Irritant

Formula: C₈H₈BrNO₄S
Melting Pt: 195…198 °C
MDL Number: MFCD01935943
CAS Number: 13029-73-7

Order Now

Specification Test Results

Melting point 195-198°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR