2-{[(4-Chlorophenyl)sulfonyl]amino}acetic acid ≥97%

Supplier: Matrix Scientific

043219-500MG 043219-1G 043219-5G
101859-738EA 81.54 USD
101859-738 101859-740 101859-742
2-{[(4-Chlorophenyl)sulfonyl]amino}acetic acid ≥97%
2-{[(4-Chlorophenyl)sulfonyl]amino}acetic acid

Hazard: Irritant

Formula: C₈H₈ClNO₄S
Melting Pt: 180…182 °C
MDL Number: MFCD01104937
CAS Number: 13029-72-6

Order Now

Specification Test Results

Melting point 180-182°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR