2-(4-Chlorophenyl)-4-thiazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

043214-500MG 043214-1G 043214-5G
101859-722EA 64.62 USD
101859-722 101859-724 101859-726
2-(4-Chlorophenyl)-4-thiazolecarboxylic acid ≥95%
2-(4-Chlorophenyl)-4-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₆NO₂ClS
Melting Pt: 187-190°C
MDL Number: MFCD00142032
CAS Number: 17228-98-7

Order Now

Specification Test Results

Melting point 187-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR