4-Methyl-2-phenyl-1,3-thiazole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methyl-2-phenylthiazole-5-carboxylic acid

043211-500MG 043211-1G 043211-5G
101859-704EA 20 USD
101859-704 101859-706 101859-708
4-Methyl-2-phenyl-1,3-thiazole-5-carboxylic acid ≥95%
4-Methyl-2-phenyl-1,3-thiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉NO₂S
MW: 219.26 g/mol
Melting Pt: 210…212 °C
MDL Number: MFCD00141955
CAS Number: 33763-20-1

Order Now

Specification Test Results

Melting point 210-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR