2-Phenyl-4-thiazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Phenylthiazole-4-carboxylic acid

043210-500MG 043210-1G 043210-5G
101859-698EA 27.69 USD
101859-698 101859-700 101859-702
2-Phenyl-4-thiazolecarboxylic acid ≥95%
2-Phenyl-4-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₇NO₂S
MW: 205.23 g/mol
Melting Pt: 210°C
MDL Number: MFCD00141954
CAS Number: 7113-10-2

Order Now

Specification Test Results

Melting point 175-177°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR