4-Chloro-1-methyl[1,2,4]triazolo[4,3-a]quinoxaline ≥95%
Supplier: Matrix Scientific
043206-500MG
043206-1G
043206-5G
101859-674EA
307.69
USD
101859-674
101859-676
101859-678
4-Chloro-1-methyl[1,2,4]triazolo[4,3-a]quinoxaline ≥95%
4-Chloro-1-methyl[1,2,4]triazolo[4,3-a]quinoxaline
Hazard: Irritant
Formula:
C₁₀H₇ClN₄ MW: 218.65 g/mol Melting Pt: 222…224 °C |
MDL Number:
MFCD00140733 CAS Number: 91895-39-5 |
Specification Test Results
Melting point | 222-224°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...