2-[(Ammoniooxy)methyl]-1-chloro-3-fluorobenzene chloride ≥95%
Supplier: Matrix Scientific
Synonyms:
O-(2-Chloro-6-fluorobenzyl)hydroxylamine hydrochloride
043205-500MG
043205-1G
043205-5G
101859-668EA
84.62
USD
101859-668
101859-670
101859-672
2-[(Ammoniooxy)methyl]-1-chloro-3-fluorobenzene chloride ≥95%
2-[(Ammoniooxy)methyl]-1-chloro-3-fluorobenzene chloride
Hazard: Irritant
Formula:
C₇H₈Cl₂FNO Melting Pt: 184-188°C |
MDL Number:
MFCD01114572 CAS Number: 93081-15-3 |
Specification Test Results
Melting point | 184-188°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...