1-[(Ammoniooxy)methyl]-3-(trifluoromethyl)benzene chloride ≥95%
Supplier: Matrix Scientific
043204-500MG
043204-1G
043204-5G
101859-662EA
366.15
USD
101859-662
101859-664
101859-666
1-[(Ammoniooxy)methyl]-3-(trifluoromethyl)benzene chloride ≥95%
1-[(Ammoniooxy)methyl]-3-(trifluoromethyl)benzene chloride
Hazard: Irritant
Formula:
C₈H₉ClF₃NO Melting Pt: 160-170°C |
MDL Number:
MFCD01114571 CAS Number: 15256-07-2 |
Specification Test Results
Melting point | 160-170°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...