1-[(Ammoniooxy)methyl]-4-methylbenzene chloride ≥95%

Supplier: Matrix Scientific

Synonyms: O-[(4-Methylphenyl)methyl]hydroxylamine hydrochloride

043203-500MG 043203-1G 043203-5G
101859-656EA 278.46 USD
101859-656 101859-658 101859-660
1-[(Ammoniooxy)methyl]-4-methylbenzene chloride ≥95%
1-[(Ammoniooxy)methyl]-4-methylbenzene chloride

Hazard: Irritant

Formula: C₈H₁₂ClNO
Melting Pt: 220-230°C
MDL Number: MFCD01114570
CAS Number: 38936-62-8

Order Now

Specification Test Results

Melting point 220-230°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR