Methyl-5-(2-methoxy-2-oxoethyl)-1-(4-methoxy-phenyl)-1H-1,2,3-triazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

043198-500MG 043198-1G 043198-5G
101859-630EA 375.38 USD
101859-630 101859-632 101859-634
Methyl-5-(2-methoxy-2-oxoethyl)-1-(4-methoxy-phenyl)-1H-1,2,3-triazole-4-carboxylate ≥95%
Methyl-5-(2-methoxy-2-oxoethyl)-1-(4-methoxy-phenyl)-1H-1,2,3-triazole-4-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₅N₃O₅
Melting Pt: 80…82 °C
MDL Number: MFCD00138716
CAS Number: 91306-62-6

Order Now

Specification Test Results

Melting point 80-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR