1-Phenyl-1H-1,2,4-triazol-3-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-1-phenyl-1H-1,2,4-triazole

043197-1G 043197-5G 043197-25G
101859-624EA 89.23 USD
101859-624 101859-626 101859-628
1-Phenyl-1H-1,2,4-triazol-3-ol ≥95%
1-Phenyl-1H-1,2,4-triazol-3-ol

Hazard: Irritant

Formula: C₈H₇N₃O
MW: 161.16 g/mol
Melting Pt: 277…280 °C
MDL Number: MFCD00138713
CAS Number: 4231-68-9

Order Now

Specification Test Results

Melting point 277-280°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR