2,2,2-Trichloro-1-[4-(4-chlorobenzoyl)-1H-pyrrol-2-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific

043195-500MG 043195-1G 043195-5G
101859-614EA 556.92 USD
101859-614 101859-616 101859-618
2,2,2-Trichloro-1-[4-(4-chlorobenzoyl)-1H-pyrrol-2-yl]-1-ethanone ≥95%
2,2,2-Trichloro-1-[4-(4-chlorobenzoyl)-1H-pyrrol-2-yl]-1-ethanone

Hazard: Irritant

Formula: C₁₃H₇Cl₄NO₂
Melting Pt: 195…197 °C
MDL Number: MFCD00141842
CAS Number: 303995-90-6

Order Now

Specification Test Results

Melting point 195-197°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR