1-[5-(2,2,2-Trichloroacetyl)-1H-pyrrol-3-yl]-1-propanone ≥95%

Supplier: Matrix Scientific

043194-500MG 043194-1G 043194-5G
101859-608EA 375.38 USD
101859-608 101859-610 101859-612
1-[5-(2,2,2-Trichloroacetyl)-1H-pyrrol-3-yl]-1-propanone ≥95%
1-[5-(2,2,2-Trichloroacetyl)-1H-pyrrol-3-yl]-1-propanone

Hazard: Irritant

Formula: C₉H₈Cl₃NO₂
Melting Pt: 190…192 °C
MDL Number: MFCD00141837
CAS Number: 111468-90-7

Order Now

Specification Test Results

Melting point 190-192°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR