1-[1,1'-Biphenyl]-4-yl-3-(dimethylamino)-2-propen-1-one ≥95%
Supplier: Matrix Scientific
043192-500MG
043192-1G
043192-5G
101859-596EA
153.85
USD
101859-596
101859-598
101859-600
1-[1,1'-Biphenyl]-4-yl-3-(dimethylamino)-2-propen-1-one ≥95%
1-[1,1'-Biphenyl]-4-yl-3-(dimethylamino)-2-propen-1-one
Hazard: Irritant
Formula:
C₁₇H₁₇NO Melting Pt: 134…136 °C |
MDL Number:
MFCD00172114 CAS Number: 138716-22-0 |
Specification Test Results
Melting point | 134-136°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...