4-(1H-Imidazol-1-yl)benzonitrile ≥95%

Supplier: Matrix Scientific

043188-500MG 043188-1G 043188-5G
101859-580EA 181.54 USD
101859-580 101859-582 101859-584
4-(1H-Imidazol-1-yl)benzonitrile ≥95%
4-(1H-Imidazol-1-yl)benzonitrile

Hazard: Irritant

Formula: C₁₀H₇N₃
MW: 169.1800 g/mol
Melting Pt: 151…153 °C
MDL Number: MFCD00127147
CAS Number: 25372-03-6

Order Now

Specification Test Results

Melting point 151-153°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR