3-Methyl-3,4,7,8-tetrahydro-2,5(1H,6H)-quinolinedione ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Methyl-1,2,3,4,5,6,7,8-octahydroquinoline-2,5-dione
043187-500MG
043187-1G
101859-576EA
278.46
USD
101859-576
101859-578
3-Methyl-3,4,7,8-tetrahydro-2,5(1H,6H)-quinolinedione ≥95%
3-Methyl-3,4,7,8-tetrahydro-2,5(1H,6H)-quinolinedione
Hazard: Irritant
Formula:
C₁₀H₁₃NO₂ MW: 179.22 g/mol Melting Pt: 206…208 °C |
MDL Number:
MFCD13184135 CAS Number: 77903-18-5 |
Specification Test Results
Melting point | 206-208°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...