1-(2-Amino-5-piperidinophenyl)-1-ethanone ≥95%
Supplier: Matrix Scientific
043186-500MG
043186-1G
043186-5G
101859-570EA
921.54
USD
101859-570
101859-572
101859-574
1-(2-Amino-5-piperidinophenyl)-1-ethanone ≥95%
1-(2-Amino-5-piperidinophenyl)-1-ethanone
Hazard: Irritant
Formula:
C₁₃H₁₈N₂O MW: 218.3 g/mol Melting Pt: 66…68 °C |
MDL Number:
MFCD08276934 CAS Number: 60283-14-9 |
Specification Test Results
Melting point | 66-68°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...