tert-Butyl-4-(3-acetyl-4-nitrophenyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(3-Acetyl-4-nitrophenyl)piperazine, N1-BOC protected

043184-500MG 043184-1G
101859-560EA 747.69 USD
101859-560 101859-562
tert-Butyl-4-(3-acetyl-4-nitrophenyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-(3-acetyl-4-nitrophenyl)tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₇H₂₃N₃O₅
MW: 349.39 g/mol
Melting Pt: 167…169 °C
MDL Number: MFCD08276930
CAS Number: 886361-04-2

Order Now

Specification Test Results

Melting point 167-169°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR