4-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
043182-500MG
043182-1G
043182-5G
101859-550EA
109.23
USD
101859-550
101859-552
101859-554
4-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid ≥95%
4-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid
Hazard: Irritant/Refrigerate
Formula:
C₆H₆BrNO₂ Melting Pt: 136…138 °C |
MDL Number:
MFCD07838483 CAS Number: 875160-43-3 |
Specification Test Results
Melting point | 136-138°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...