2-Methyl-1,3-benzoxazol-4-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Amino-2-methyl-1,3-benzoxazole
043181-500MG
043181-1G
043181-5G
101859-544EA
153.85
USD
101859-544
101859-546
101859-548
2-Methyl-1,3-benzoxazol-4-amine ≥95%
2-Methyl-1,3-benzoxazol-4-amine
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O Melting Pt: 76…78 °C |
CAS Number:
342897-54-5 |
Specification Test Results
Melting point | 76-78°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...