Methyl-5-phenylisoxazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-phenylisoxazole-3-carboxylate

043176-500MG 043176-1G 043176-5G
101859-516EA 278.46 USD
101859-516 101859-518 101859-520
Methyl-5-phenylisoxazole-3-carboxylate ≥95%
Methyl-5-phenylisoxazole-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₉NO₃
MW: 203.1941 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD02159553
CAS Number: 51677-09-9

Order Now

Specification Test Results

Melting point 82-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR