4-[4-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
043175-500MG
043175-1G
101859-512EA
375.38
USD
101859-512
101859-514
4-[4-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde ≥95%
4-[4-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₃H₈F₃NO MW: 251.21 g/mol Melting Pt: 66…68 °C |
MDL Number:
MFCD09607891 CAS Number: 952182-74-0 |
Specification Test Results
Melting point | 66-68°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...