2-Amino-1-(4-bromophenyl)-1-ethanol ≥95%
Supplier: Matrix Scientific
Synonyms:
alpha-(Aminomethyl)-4-bromobenzyl alcohol
043174-500MG
043174-1G
043174-5G
101859-506EA
223.08
USD
101859-506
101859-508
101859-510
2-Amino-1-(4-bromophenyl)-1-ethanol ≥95%
2-Amino-1-(4-bromophenyl)-1-ethanol
Hazard: Irritant
Formula:
C₈H₁₀BrNO MW: 216.08 g/mol Melting Pt: 111…113 °C |
MDL Number:
MFCD08276931 CAS Number: 41147-82-4 |
Specification Test Results
Melting point | 111-113°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...