1-Boc-3-hydroxymethyl-5-methoxyindole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Hydroxymethyl)-5-methoxy-1H-indole, N-BOC protected

043173-500MG 043173-1G 043173-5G
101859-500EA 375.38 USD
101859-500 101859-502 101859-504
1-Boc-3-hydroxymethyl-5-methoxyindole ≥95%
1-Boc-3-hydroxymethyl-5-methoxyindole

Hazard: Irritant

Formula: C₁₅H₁₉NO₄
Melting Pt: 137…139 °C
CAS Number: 600136-09-2

Order Now

Specification Test Results

Melting point 137-139°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR