4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

043171-500MG 043171-1G
101859-490EA 278.46 USD
101859-490 101859-492
4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde ≥95%
4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₇ClF₃NO
MW: 285.66 g/mol
Melting Pt: 71…73 °C
MDL Number: MFCD09607889
CAS Number: 952182-72-8

Order Now

Specification Test Results

Melting point 71-73°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR