[2-(4-Methoxyphenyl)-1,3-thiazol-4-yl]methanol ≥95%
Supplier: Matrix Scientific
043169-500MG
043169-1G
101859-486EA
747.69
USD
101859-486
101859-488
[2-(4-Methoxyphenyl)-1,3-thiazol-4-yl]methanol ≥95%
[2-(4-Methoxyphenyl)-1,3-thiazol-4-yl]methanol
Hazard: Irritant
Formula:
C₁₁H₁₁NO₂S Melting Pt: 84…86 °C |
MDL Number:
MFCD06738392 CAS Number: 885279-75-4 |
Specification Test Results
Melting point | 84-86°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...