2-[4-(tert-Butyl)phenyl]-5-(chloromethyl)-1,3,4-oxadiazole ≥95%
Supplier: Matrix Scientific
043164-500MG
043164-1G
101859-470EA
747.69
USD
101859-470
101859-472
2-[4-(tert-Butyl)phenyl]-5-(chloromethyl)-1,3,4-oxadiazole ≥95%
2-[4-(tert-Butyl)phenyl]-5-(chloromethyl)-1,3,4-oxadiazole
Hazard: Irritant
Formula:
C₁₃H₁₅ClN₂O Melting Pt: 69…70 °C |
MDL Number:
MFCD06797381 CAS Number: 754214-36-3 |
Specification Test Results
Melting point | 69-70°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...