2-(Chloromethyl)-5-[4-(trifluoromethyl)phenyl]-1,3,4-oxadiazole ≥95%

Supplier: Matrix Scientific

043163-500MG 043163-1G
101859-466EA 921.54 USD
101859-466 101859-468
2-(Chloromethyl)-5-[4-(trifluoromethyl)phenyl]-1,3,4-oxadiazole ≥95%
2-(Chloromethyl)-5-[4-(trifluoromethyl)phenyl]-1,3,4-oxadiazole

Hazard: Irritant

Formula: C₁₀H₆ClF₃N₂O
Melting Pt: 79…81 °C
MDL Number: MFCD06797380
CAS Number: 753479-67-3

Order Now

Specification Test Results

Melting point 79-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR