2-(Chloromethyl)-5-(4-chlorophenyl)-1,3,4-oxadiazole ≥95%
Supplier: Matrix Scientific
043162-500MG
043162-1G
043162-5G
101859-460EA
556.92
USD
101859-460
101859-462
101859-464
2-(Chloromethyl)-5-(4-chlorophenyl)-1,3,4-oxadiazole ≥95%
2-(Chloromethyl)-5-(4-chlorophenyl)-1,3,4-oxadiazole
Hazard: Irritant
Formula:
C₉H₆Cl₂N₂O Melting Pt: 83…85 °C |
MDL Number:
MFCD00714902 CAS Number: 24068-15-3 |
Specification Test Results
Melting point | 83-85°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...