Methyl-2-amino-5-bromo-3-methylbenzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-amino-5-bromo-3-methylbenzoate
043160-500MG
043160-1G
043160-5G
101859-450EA
747.69
USD
101859-450
101859-452
101859-454
Methyl-2-amino-5-bromo-3-methylbenzenecarboxylate ≥95%
Methyl-2-amino-5-bromo-3-methylbenzenecarboxylate
Hazard: Irritant
Formula:
C₉H₁₀BrNO₂ MW: 244.09 g/mol Melting Pt: 56…58 °C |
MDL Number:
MFCD06797379 CAS Number: 206548-14-3 |
Specification Test Results
Melting point | 56-58°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...