N-Methyl-N-(3-nitrophenyl)acetamide ≥95%
Supplier: Matrix Scientific
043159-500MG
043159-1G
043159-5G
101859-444EA
181.54
USD
101859-444
101859-446
101859-448
N-Methyl-N-(3-nitrophenyl)acetamide ≥95%
N-Methyl-N-(3-nitrophenyl)acetamide
Hazard: Irritant
Formula:
C₉H₁₀N₂O₃ Melting Pt: 99…101 °C |
MDL Number:
MFCD02169677 CAS Number: 21353-89-9 |
Specification Test Results
Melting point | 99-101°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...