2-(4-Chlorophenyl)-1-(1-ethylpropyl)-6-oxo-3-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

043158-500MG 043158-1G 043158-5G
101859-438EA 556.92 USD
101859-438 101859-440 101859-442
2-(4-Chlorophenyl)-1-(1-ethylpropyl)-6-oxo-3-piperidinecarboxylic acid ≥95%
2-(4-Chlorophenyl)-1-(1-ethylpropyl)-6-oxo-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₂ClNO₃
Melting Pt: 213…215 °C
MDL Number: MFCD08056606
CAS Number: 886360-97-0

Order Now

Specification Test Results

Melting point 213-215°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR