tert-Butyl-1-oxa-4,8-diazaspiro[4.5]decane-8-carboxylate ≥95%
Supplier: Matrix Scientific
043157-500MG
043157-1G
043157-5G
101859-432EA
278.46
USD
101859-432
101859-434
101859-436
tert-Butyl-1-oxa-4,8-diazaspiro[4.5]decane-8-carboxylate ≥95%
tert-Butyl-1-oxa-4,8-diazaspiro[4.5]decane-8-carboxylate
Hazard: Irritant
Formula:
C₁₂H₂₂N₂O₃ Melting Pt: 48…50 °C |
MDL Number:
MFCD07698579 CAS Number: 886360-95-8 |
Specification Test Results
Melting point | 48-50°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...