Benzyl-3-oxopiperazine-1-carboxylate ≥97%

Supplier: Matrix Scientific

043155-1G 043155-5G
101859-424EA 76.92 USD
101859-424 102551-316
Benzyl-3-oxopiperazine-1-carboxylate ≥97%
Benzyl-3-oxopiperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₃
Melting Pt: 107…110 °C
MDL Number: MFCD00173924
CAS Number: 78818-15-2

Order Now

Specification Test Results

Melting point 107-110°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR