tert-Butyl-N-[3-(2-methyl-1,3-dioxolan-2-yl)phenyl]carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(2-Methyl-1,3-dioxolan-2-yl)aniline, N-BOC protected

043154-500MG 043154-1G 043154-5G
101859-416EA 556.92 USD
101859-416 101859-418 101859-420
tert-Butyl-N-[3-(2-methyl-1,3-dioxolan-2-yl)phenyl]carbamate ≥95%
tert-Butyl-N-[3-(2-methyl-1,3-dioxolan-2-yl)phenyl]carbamate

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
Melting Pt: 63 °C
MDL Number: MFCD06200926
CAS Number: 886361-42-8

Order Now

Specification Test Results

Melting point 63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR