Methyl-2-amino-5-(2-fluorophenyl)-1,3-thiazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
043151-500MG
043151-1G
043151-5G
101859-404EA
90.77
USD
101859-404
101859-406
101859-408
Methyl-2-amino-5-(2-fluorophenyl)-1,3-thiazole-4-carboxylate ≥95%
Methyl-2-amino-5-(2-fluorophenyl)-1,3-thiazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₁H₉FN₂O₂S MW: 252.27 g/mol Melting Pt: 203 °C |
MDL Number:
MFCD06200924 CAS Number: 886361-38-2 |
Specification Test Results
Melting point | 203°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...