5-Amino-1-(4-methylphenyl)-1H-pyrazole-4-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-1-(p-tolyl)-1H-pyrazole-4-carbonitrile, 5-Amino-1-(4-methylphenyl)pyrazole-4-carbonitrile, 5-Amino-1-(p-tolyl)pyrazole-4-carbonitrile

043150-500MG 043150-1G 043150-5G
101859-398EA 181.54 USD
101859-398 101859-400 101859-402
5-Amino-1-(4-methylphenyl)-1H-pyrazole-4-carbonitrile ≥95%
5-Amino-1-(4-methylphenyl)-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₁₁H₁₀N₄
MW: 198.23 g/mol
Melting Pt: 116…120 °C
MDL Number: MFCD00052944
CAS Number: 103646-82-8

Order Now

Specification Test Results

Melting point 116-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR