Methyl-2-amino-5-(4-cyanophenyl)-1,3-thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

043149-500MG 043149-1G 043149-5G
101859-392EA 124.62 USD
101859-392 101859-394 101859-396
Methyl-2-amino-5-(4-cyanophenyl)-1,3-thiazole-4-carboxylate ≥95%
Methyl-2-amino-5-(4-cyanophenyl)-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₉N₃O₂S
MW: 259.29 g/mol
Melting Pt: 266…268 °C
MDL Number: MFCD06200923
CAS Number: 886361-36-0

Order Now

Specification Test Results

Melting point 266-268°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR