Methyl-2-amino-5-(3-fluorophenyl)-1,3-thiazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
043148-500MG
043148-1G
043148-5G
101859-386EA
181.54
USD
101859-386
101859-388
101859-390
Methyl-2-amino-5-(3-fluorophenyl)-1,3-thiazole-4-carboxylate ≥95%
Methyl-2-amino-5-(3-fluorophenyl)-1,3-thiazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₁H₉FN₂O₂S MW: 252.27 g/mol Melting Pt: 216 °C |
MDL Number:
MFCD06200922 CAS Number: 127918-93-8 |
Specification Test Results
Melting point | 216°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...