Methyl-2-amino-5-[4-(tert-butyl)phenyl]-1,3-thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

043147-500MG 043147-1G 043147-5G
101859-380EA 181.54 USD
101859-380 101859-382 101859-384
Methyl-2-amino-5-[4-(tert-butyl)phenyl]-1,3-thiazole-4-carboxylate ≥95%
Methyl-2-amino-5-[4-(tert-butyl)phenyl]-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₂S
MW: 290.39 g/mol
Melting Pt: 232…234 °C
MDL Number: MFCD06200921
CAS Number: 886361-34-8

Order Now

Specification Test Results

Melting point 232-234°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR