Methyl-4-amino-3-methylbenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-amino-3-methylbenzoate

043141-25G 043141-100G
101859-372EA 29.23 USD
101859-372 102833-120
Methyl-4-amino-3-methylbenzoate ≥95%
Methyl 4-amino-3-methylbenzoate

Hazard: Irritant

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 117-119°C
MDL Number: MFCD00102230
CAS Number: 18595-14-7

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR