Methyl quinoxaline-6-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 6-Quinoxalinecarboxylate

043139-1G 043139-5G
101859-366EA 186.15 USD
101859-366 101859-368
Methyl quinoxaline-6-carboxylate ≥95%
Methyl quinoxaline-6-carboxylate

Hazard: Irritant

Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
Melting Pt: 97-99°C
MDL Number: MFCD00102618
CAS Number: 23088-23-5

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR