5-(4-Bromophenyl)oxazole ≥95%
Supplier: Matrix Scientific
043137-500MG
043137-5G
101859-354EA
60
USD
101859-354
101859-358
5-(4-Bromophenyl)oxazole ≥95%
5-(4-Bromophenyl)oxazole
Hazard: Irritant
Formula:
C₉H₆BrNO MW: 224.0540 g/mol Melting Pt: 78-80°C |
MDL Number:
MFCD02682015 CAS Number: 72571-06-3 |
Specification Test Results
Melting point | 78-80°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...